2-(4-Methoxyphenyl)benzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Benzothiazolyl)anisole, 1-(2-Benzothiazolyl)-4-methoxybenzene

A386114-1G A386114-25G A386114-5G
77142-648EA 17.52 USD
77142-648 77142-650 77142-652
2-(4-Methoxyphenyl)benzothiazole 98%
2-(4-Methoxyphenyl)benzothiazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁NOS
MDL Number: MFCD01099069
CAS Number: 6265-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR