(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2-methylpyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-α-Me-Pro-OH

A384938-100MG A384938-1G A384938-250MG A384938-5G
77142-630EA 28.83 USD
77142-630 77142-632 77142-634 77746-774
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2-methylpyrrolidine-2-carboxylic acid 97%
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-2-methylpyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD03095629
CAS Number: 167275-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR