4-Bromo-4',4''-dimethyltriphenylamine 99%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-4,4-dimethyltriphenylamine, 4-Bromo-N,N-di-p-tolylaniline

A381464-100G A381464-1G A381464-5G
77142-606EA 299.18 USD
77142-606 77142-608 77142-610
4-Bromo-4',4''-dimethyltriphenylamine 99%
4-Bromo-4',4''-dimethyltriphenylamine

Precaución: for Research Use Only

Formula: C₂₀H₁₈BrN
MW: 352.28 g/mol
MDL Number: MFCD03093258
CAS Number: 58047-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR