(1-Hydroxyethylidene)bis-phosphonic acid tetrasodium salt 98%

Supplier: Ambeed
Warning

A379605-100G A379605-25G A379605-500G
77142-586EA 37.08 USD
77142-586 77142-588 77142-590
(1-Hydroxyethylidene)bis-phosphonic acid tetrasodium salt 98%
(1-Hydroxyethylidene)bis-phosphonic acid tetrasodium salt

Precaución: for Research Use Only

Formula: C₂H₄Na₄O₇P₂
MDL Number: MFCD01729922
CAS Number: 3794-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR