4-Chlorophenyl benzoate 97%

Supplier: Ambeed
Warning

A379579-1G A379579-25G A379579-5G
77142-580EA 13.39 USD
77142-580 77142-582 77142-584
4-Chlorophenyl benzoate 97%
4-Chlorophenyl benzoate

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
MDL Number: MFCD00040854
CAS Number: 2005-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR