(4R,12aS)-7-Methoxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxylic acid 97%

Supplier: Ambeed
Warning

A378556-100MG A378556-1G
77142-564EA 6.18 USD
77142-564 77746-760
(4R,12aS)-7-Methoxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxylic acid 97%
(4R,12aS)-7-Methoxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₆
MDL Number: MFCD28978321
CAS Number: 1335210-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR