2,4-Dimethyl-3-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A377872-100MG A377872-250MG
77142-556EA 43.25 USD
77142-556 77142-558
2,4-Dimethyl-3-nitrobenzonitrile 97%
2,4-Dimethyl-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD08693500
CAS Number: 39053-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR