(R)-3-N-Cbz-Aminopyrrolidine 95%

Supplier: Ambeed
Warning

A376227-250MG A376227-5G
77142-536EA 18.04 USD
77142-536 77142-538
(R)-3-N-Cbz-Aminopyrrolidine 95%
(R)-3-N-Cbz-Aminopyrrolidine

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MW: 220.26 g/mol
MDL Number: MFCD04115316
CAS Number: 879275-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR