Methyl 2-amino-3,5-difluorobenzoate 97%
Supplier: Ambeed
|
Warning
|
A375435-1G
A375435-250MG
77142-524EA
36.05
USD
77142-524
77142-526
Methyl 2-amino-3,5-difluorobenzoate 97%
Methyl 2-amino-3,5-difluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₇F₂NO₂ |
MDL Number:
MFCD12159866 CAS Number: 1184559-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...