1-(4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A375269-1G A375269-250MG A375269-5G
77142-516EA 133.68 USD
77142-516 77142-518 77142-520
1-(4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride 97%
1-(4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₄BClFNO₂
MW: 315.61 g/mol
MDL Number: MFCD20529511
CAS Number: 2096996-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR