1,2-Dibromo-4,5-diethoxy-3-fluorobenzene 97%

Supplier: Ambeed
Warning

A373664-10G A373664-1G A373664-5G
77142-500EA 249.32 USD
77142-500 77142-502 77142-504
1,2-Dibromo-4,5-diethoxy-3-fluorobenzene 97%
1,2-Dibromo-4,5-diethoxy-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁Br₂FO₂
MW: 342.00 g/mol
MDL Number: MFCD22050880
CAS Number: 474554-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR