4-Ethoxy-2,3-difluoro-6-methylphenol 97%

Supplier: Ambeed
Warning

A372766-100MG A372766-1G A372766-250MG
77142-494EA 65.78 USD
77142-494 77142-496 77142-498
4-Ethoxy-2,3-difluoro-6-methylphenol 97%
4-Ethoxy-2,3-difluoro-6-methylphenol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
MW: 188.17 g/mol
MDL Number: MFCD28291820
CAS Number: 886037-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR