Dicarbonyl(2,4-pentanedionato)iridium(I) 98%

Supplier: Ambeed
Danger

Synonyms: (Acetylacetonato)dicarbonyliridium(I), Dicarbonyl(acetylacetonato)iridium(I)

A370583-100MG A370583-1G A370583-250MG A370583-5G
77142-466EA 46.68 USD
77142-466 77142-468 77142-470 77142-472
Dicarbonyl(2,4-pentanedionato)iridium(I) 98%
Dicarbonyl(2,4-pentanedionato)iridium(I)

Precaución: for Research Use Only

Formula: C₇H₇IrO₄
MW: 347.35 g/mol
MDL Number: MFCD00049135
CAS Number: 14023-80-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR