N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic acid

A370315-100MG A370315-1G A370315-250MG A370315-5G
77142-450EA 64.72 USD
77142-450 77142-452 77142-454 77142-456
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MW: 333.303 g/mol
MDL Number: MFCD06659150
CAS Number: 922178-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR