(Iodomethyl)cyclopropane ≥95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A369443-1G A369443-250MG A369443-25G A369443-5G
77142-428EA 33.95 USD
77142-428 77142-430 77142-432 77568-754
(Iodomethyl)cyclopropane ≥95% stabilized
(Iodomethyl)cyclopropane

Precaución: For research use only.

Formula: C₄H₇I
MW: 182.00 g/mol
MDL Number: MFCD06410734
CAS Number: 33574-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR