Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')manganese(III) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Mn(dpm)3, Mn(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)manganese(III)
A368026-1G
A368026-25G
A368026-5G
A368026-100G
77142-414EA
9.26
USD
77142-414
77142-416
77142-418
77616-610
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')manganese(III) 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')manganese(III)
Precaución: for Research Use Only
Formula:
C₃₃H₅₇MnO₆ |
MDL Number:
MFCD00269825 CAS Number: 14324-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...