Z-Trp-OMe 95%

Supplier: Ambeed
Warning

A367173-100G A367173-10G A367173-1G A367173-250MG
77142-394EA 180.24 USD
77142-394 77142-396 77142-398 77142-400
Z-Trp-OMe 95%
Z-Trp-OMe

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₄
MDL Number: MFCD06658545
CAS Number: 2717-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR