7-Chloroquinolin-4(1H)-one 97%

Supplier: Ambeed
Warning

A366847-100G A366847-25G A366847-5G
77142-388EA 337.83 USD
77142-388 77142-390 77142-392
7-Chloroquinolin-4(1H)-one 97%
7-Chloroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD20542777
CAS Number: 23833-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR