Methyl 5-amino-2-fluoro-4-methylbenzoate 95%

Supplier: Ambeed
Warning

A365381-1G A365381-250MG
77142-380EA 41.2 USD
77142-380 77142-382
Methyl 5-amino-2-fluoro-4-methylbenzoate 95%
Methyl 5-amino-2-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD23863661
CAS Number: 1504965-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR