2-Formyl-6-methoxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxypicolinaldehyde, 6-Methoxypyridine-2-carboxaldehyde, 6-Methoxypyridine-2-carbaldehyde

A364712-10G A364712-1G A364712-25G A364712-5G
77142-362EA 54.11 USD
77142-362 77142-364 77142-366 77142-368
2-Formyl-6-methoxypyridine 97%
2-Formyl-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD05664030
CAS Number: 54221-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR