Tiplaxtinin 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
PAI 039, Tiplaxtinin, 2-[1-benzyl-5-[4-(trifluoromethoxy)phenyl]indol-3-yl]-2-oxoacetic acid, PAI039, PAI-039, 2-(1-Benzyl-5-(4-(trifluoromethoxy)phenyl)-1H-indol-3-yl)-2-oxoacetic acid, Tiplasinin
A363712-10MG
A363712-1MG
A363712-25MG
A363712-50MG
A363712-5MG
77142-352EA
132.62
USD
77142-352
77142-354
77142-356
77142-358
77142-360
Tiplaxtinin 98%
Tiplaxtinin
Precaución: for Research Use Only
Formula:
C₂₄H₁₆F₃NO₄ MW: 439.38 g/mol |
MDL Number:
MFCD09475615 CAS Number: 393105-53-8 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...