2,2'-Bipyridinedichloroplatinum 96% Pt Assay:46.21 ± 1.00%

Supplier: Ambeed
Warning

A363696-100MG A363696-1G A363696-250MG
77142-346EA 78.28 USD
77142-346 77142-348 77142-350
2,2'-Bipyridinedichloroplatinum 96% Pt Assay:46.21 ± 1.00%
2,2'-Bipyridinedichloroplatinum

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂N₂Pt
MDL Number: MFCD00191790
CAS Number: 13965-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR