2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A362768-1G A362768-250MG A362768-5G A362768-100MG
77142-330EA 18.55 USD
77142-330 77142-332 77142-334 77686-026
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₂₃H₁₉N₃O₂
MDL Number: MFCD01863585
CAS Number: 128249-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR