Methyl 3-chloro-4-methylthiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

A361302-10G A361302-1G A361302-5G
77142-322EA 98.67 USD
77142-322 77142-324 77746-719
Methyl 3-chloro-4-methylthiophene-2-carboxylate 98%
Methyl 3-chloro-4-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClO₂S
MW: 190.647 g/mol
MDL Number: MFCD00052072
CAS Number: 175137-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR