(+)-Dipivaloyl-D-tartaric acid 98%

Supplier: Ambeed
Warning

Synonyms: (+)-Bis(2,2-dimethylpropionyl)-D-tartaric Acid, (2S,3S)-2,3-Bis(pivaloyloxy)succinic acid, (+)-O,O'-Dipivaloyl-D-tartaric acid

A361040-1G A361040-25G A361040-5G
77142-310EA 13.39 USD
77142-310 77142-312 77142-314
(+)-Dipivaloyl-D-tartaric acid 98%
(+)-Dipivaloyl-D-tartaric acid

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₈
MDL Number: MFCD00015634
CAS Number: 76769-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR