4-Bromo-2,6-dimethoxypyrimidine 97%

Supplier: Ambeed
Warning

A360771-100MG A360771-5G
77142-306EA 42.44 USD
77142-306 77142-308
4-Bromo-2,6-dimethoxypyrimidine 97%
4-Bromo-2,6-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.03 g/mol
MDL Number: MFCD07787454
CAS Number: 60186-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR