4-Chloro-2-(trifluoromethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane

A360190-10G A360190-1G A360190-250MG A360190-25G A360190-5G
77142-294EA 43.5 USD
77142-294 77142-296 77142-298 77142-300 77142-302
4-Chloro-2-(trifluoromethyl)phenylboronic acid 97%
4-Chloro-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD04973086
CAS Number: 313545-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR