4-(2-Fluoro-3-nitrophenyl)pyridine 95%

Supplier: Ambeed
Warning

A359942-100MG A359942-250MG
77142-286EA 78.51 USD
77142-286 77142-288
4-(2-Fluoro-3-nitrophenyl)pyridine 95%
4-(2-Fluoro-3-nitrophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇FN₂O₂
MW: 218.18 g/mol
MDL Number: MFCD15475666
CAS Number: 1214391-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR