(S)-(-)-2-(Boc-amino)-1,4-butanediol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(tert-butoxycarbonylamino)-1,4-butanediol, (S)-N-Boc-2-Aminobutane-1,4-diol, tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate

A359831-1G A359831-250MG A359831-5G
77142-274EA 21.22 USD
77142-274 77142-276 77142-278
(S)-(-)-2-(Boc-amino)-1,4-butanediol 97%
(S)-(-)-2-(Boc-amino)-1,4-butanediol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₄
MW: 205.254 g/mol
MDL Number: MFCD03093946
CAS Number: 128427-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR