(S)-(+)-α-Cyclohexanemethylamine 98%

Supplier: Ambeed
Danger

Synonyms: (S)-1-Cyclohexylethanamine, (S)-(+)-1-Cyclohexylethylamine

A359348-1G A359348-25G A359348-5G
77142-268EA 19.57 USD
77142-268 77142-270 77142-272
(S)-(+)-α-Cyclohexanemethylamine 98%
(S)-(+)-α-Cyclohexanemethylamine

Precaución: for Research Use Only

Formula: C₈H₁₇N
MDL Number: MFCD00066261
CAS Number: 17430-98-7
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR