1-(Cyanomethyl)pyridinium chloride 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Cyanomethyl)pyridin-1-ium chloride

A358889-1G A358889-25G A358889-5G
77142-262EA 9.27 USD
77142-262 77142-264 77142-266
1-(Cyanomethyl)pyridinium chloride 98%
1-(Cyanomethyl)pyridinium chloride

Precaución: for Research Use Only

Formula: C₇H₇ClN₂
MDL Number: MFCD00031933
CAS Number: 17281-59-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR