3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A358789-100MG
A358789-1G
A358789-250MG
77142-256EA
36.08
USD
77142-256
77142-258
77142-260
3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₆BNO₄ MW: 273.09 g/mol |
MDL Number:
MFCD22586692 CAS Number: 2377611-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...