(R)-N,N-Dimethyl-1-[(S)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-(S)-BPPFA, (R)-N,N-Dimethyl-1-[(S)-1′,2-bis(diphenylphosphino)ferrocenyl]ethylamine

A357981-100MG A357981-1G A357981-250MG
77142-248EA 68.96 USD
77142-248 77142-250 77142-252
(R)-N,N-Dimethyl-1-[(S)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine ≥98%, ee 99%
(R)-N,N-Dimethyl-1-[(S)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine

Precaución: for Research Use Only

Formula: C₃₈H₃₇FeNP₂
MW: 625.51 g/mol
MDL Number: MFCD00075284
CAS Number: 74311-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR