2-Bromo-4,5-diluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-2-methylbromobenzene, 1-Bromo-3,4-difluoro-6-methylbenzene, 1-Bromo-4,5-difluoro-2-methylbenzene

A355995-10G A355995-1G A355995-250MG A355995-25G A355995-5G
77142-222EA 131.56 USD
77142-222 77142-224 77142-226 77142-228 77142-230
2-Bromo-4,5-diluorotoluene 98%
2-Bromo-4,5-diluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD07777160
CAS Number: 875664-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR