Methyl 2-(3-hydroxy-4-nitrophenyl)acetate 97%
Supplier: Ambeed
|
Warning
|
A354561-100MG
A354561-250MG
77142-216EA
23.69
USD
77142-216
77142-218
Methyl 2-(3-hydroxy-4-nitrophenyl)acetate 97%
Methyl 2-(3-hydroxy-4-nitrophenyl)acetate
Precaución: for Research Use Only
Formula:
C₉H₉NO₅ |
MDL Number:
MFCD18399396 CAS Number: 256453-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...