(R)-2-Amino-2-(thiophen-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-α-(2-Thienyl)-Gly-OH

A354433-100MG A354433-5G
77142-212EA 51.99 USD
77142-212 77142-214
(R)-2-Amino-2-(thiophen-2-yl)acetic acid 95%
(R)-2-Amino-2-(thiophen-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
MDL Number: MFCD00079612
CAS Number: 65058-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR