(2-(Benzyloxy)-4-cyanophenyl)boronic acid 97%

Supplier: Ambeed
Warning

A353767-1G A353767-250MG
77142-202EA 75.19 USD
77142-202 77142-204
(2-(Benzyloxy)-4-cyanophenyl)boronic acid 97%
(2-(Benzyloxy)-4-cyanophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BNO₃
MDL Number: MFCD20441839
CAS Number: 1884477-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR