(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A353650-1G A353650-250MG A353650-5G
77142-196EA 143.17 USD
77142-196 77142-198 77142-200
(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)boronic acid 97%
(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BF₄O₃
MDL Number: MFCD19981509
CAS Number: 2096332-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR