(Dimethylamino)acetone 98%

Supplier: Ambeed
Danger

A349647-1G A349647-250MG
77142-182EA 58.71 USD
77142-182 77142-184
(Dimethylamino)acetone 98%
(Dimethylamino)acetone

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MDL Number: MFCD00008766
CAS Number: 15364-56-4
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR