Methyl 6-bromo-5-chloropicolinate 95%

Supplier: Ambeed
Warning

A347278-100MG A347278-1G A347278-250MG
77142-162EA 94.43 USD
77142-162 77142-164 77142-166
Methyl 6-bromo-5-chloropicolinate 95%
Methyl 6-bromo-5-chloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.47 g/mol
MDL Number: MFCD14698099
CAS Number: 1214324-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR