2,2'-((Pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulfonate 98%

Supplier: Ambeed
Warning

A346096-1G A346096-25G A346096-5G
77142-156EA 76.21 USD
77142-156 77142-158 77142-160
2,2'-((Pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulfonate 98%
2,2'-((Pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulfonate

Precaución: for Research Use Only

Formula: C₆₅H₈₂N₂O₁₈S₂
MDL Number: MFCD26131271
CAS Number: 64228-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR