(2-tert-Butylphenoxy)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(tert-Butyl)phenoxy)acetic acid

A343646-100MG A343646-1G A343646-250MG A343646-5G
77142-140EA 13.8 USD
77142-140 77142-142 77142-144 77142-146
(2-tert-Butylphenoxy)acetic acid 95%
(2-tert-Butylphenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
MDL Number: MFCD06655706
CAS Number: 19271-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR