2-Chloro-6-methyl-4-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A343573-100G A343573-10G A343573-1G A343573-250MG A343573-25G A343573-5G
77142-128EA 682.16 USD
77142-128 77142-130 77142-132 77142-134 77142-136 77142-138
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine 95%
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MDL Number: MFCD00052900
CAS Number: 22123-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR