(S)-(-)-2-Methyl-1-butanol 98%

Supplier: Ambeed
Danger

A343263-25G A343263-5G
77142-124EA 10.3 USD
77142-124 77142-126
(S)-(-)-2-Methyl-1-butanol 98%
(S)-(-)-2-Methyl-1-butanol

Precaución: for Research Use Only

Formula: C₅H₁₂O
MDL Number: MFCD00064299
CAS Number: 1565-80-6
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR