(6-(3-Methoxyphenoxy)pyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A343069-1G A343069-250MG A343069-5G
77142-118EA 161.26 USD
77142-118 77142-120 77142-122
(6-(3-Methoxyphenoxy)pyridin-3-yl)boronic acid 97%
(6-(3-Methoxyphenoxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂BNO₄
MW: 245.04 g/mol
MDL Number: MFCD22375022
CAS Number: 2096338-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR