2-(6-(((Benzhydryloxy)carbonyl)amino)-9H-purin-9-yl)acetic acid 95%

Supplier: Ambeed
Warning

A341952-100MG A341952-1G A341952-250MG A341952-5G
77142-104EA 9.55 USD
77142-104 77142-106 77142-108 77142-110
2-(6-(((Benzhydryloxy)carbonyl)amino)-9H-purin-9-yl)acetic acid 95%
2-(6-(((Benzhydryloxy)carbonyl)amino)-9H-purin-9-yl)acetic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₇N₅O₄
MW: 403.39 g/mol
MDL Number: MFCD09261306
CAS Number: 186046-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR