Methyl 2-(benzylamino)acetate hydrochloride 97%

Supplier: Ambeed
Warning

A340718-1G A340718-25G A340718-5G
77142-084EA 20.6 USD
77142-084 77142-086 77142-088
Methyl 2-(benzylamino)acetate hydrochloride 97%
Methyl 2-(benzylamino)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂.HCl
MDL Number: MFCD02684379
CAS Number: 17136-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR