(1,5-Dimethyl-1H-pyrazol-3-yl)methylamine 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)-1,5-dimethylpyrazole, 3-(Aminomethyl)-1,5-dimethyl-1H-pyrazole

A338887-250MG A338887-1G
77142-062EA 44.56 USD
77142-062 77571-978
(1,5-Dimethyl-1H-pyrazol-3-yl)methylamine 97%
(1,5-Dimethyl-1H-pyrazol-3-yl)methylamine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃
MW: 125.17 g/mol
MDL Number: MFCD03422531
CAS Number: 423768-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR