1,6-Dibromo-3,8-diisopropylpyrene 95%

Supplier: Ambeed
Warning

A338225-250MG A338225-5G
77142-048EA 11.34 USD
77142-048 77685-970
1,6-Dibromo-3,8-diisopropylpyrene 95%
1,6-Dibromo-3,8-diisopropylpyrene

Precaución: for Research Use Only

Formula: C₂₂H₂₀Br₂
MDL Number: MFCD27979119
CAS Number: 869340-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR