2,4-Dibromonicotinic acid 97%

Supplier: Ambeed
Warning

A338103-100MG A338103-1G A338103-250MG A338103-50MG
77142-040EA 221.73 USD
77142-040 77142-042 77142-044 77142-046
2,4-Dibromonicotinic acid 97%
2,4-Dibromonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.90 g/mol
MDL Number: MFCD18262257
CAS Number: 1269291-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR